Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007483.hg.1 |
ENST00000358339.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007483.hg.1 |
ENST00000371485.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007483.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007483.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007483.hg.1 |
TC10000664.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000496302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000496302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000496302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
ENST00000496302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157677.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157677.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157677.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157677.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157677.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157677.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157677.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157677.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157677.hg.1 |
TC1000008482.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000371485.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000445435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000496302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000496302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000496302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
ENST00000496302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_001127182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008482.hg.1 |
NM_018131.4 |
Refseq |