Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023053.hg.1 |
TC06001896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023053.hg.1 |
ENST00000275036.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023053.hg.1 |
ENST00000344726.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023053.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023053.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023053.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023053.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023057.hg.1 |
TC06001896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023057.hg.1 |
ENST00000344726.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023057.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023057.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023057.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185227.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185227.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185227.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185227.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185227.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185227.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185227.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185227.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185227.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185227.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185227.hg.1 |
TC0600012363.hg.1 |
Clariom D TC |