Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13010114.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13010114.hg.1 |
ENST00000389544.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13010114.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13010114.hg.1 |
TC13000901.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000542651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000542651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000542651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000542651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000542651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
ENST00000542651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000542651.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000542651.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000542651.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000542651.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000542651.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
ENST00000542651.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009933.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166142.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166142.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166142.hg.1 |
ENST00000334062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166142.hg.1 |
NM_007368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166142.hg.1 |
TC1300009933.hg.1 |
Clariom D TC |