Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018303.5 | NP_060773.3 | 24 - 25 | 2600 | 98 | |
NR_073064.1 | 26 - 27 | 2762 | 98 | |||
XM_017011018.1 | XP_016866507.1 | 25 - 26 | 2646 | 98 | ||
XM_017011019.1 | XP_016866508.1 | 25 - 26 | 2808 | 98 | ||
XM_017011020.1 | XP_016866509.1 | 25 - 26 | 2668 | 98 | ||
XM_017011021.1 | XP_016866510.1 | 24 - 25 | 2904 | 98 | ||
XM_017011022.1 | XP_016866511.1 | 24 - 25 | 2600 | 98 | ||
XM_017011023.1 | XP_016866512.1 | 24 - 25 | 2600 | 98 | ||
XM_017011024.1 | XP_016866513.1 | 25 - 26 | 2646 | 98 | ||
GenBank mRNA | AJ414403.1 | - | 24 - 25 | 2524 | 98 | |
AJ420556.1 | - | 2 - 3 | 119 | 98 | ||
AK001888.1 | - | 20 - 21 | 2015 | 98 | ||
AK056564.1 | - | 24 - 25 | 2568 | 98 | ||
AK302941.1 | - | 14 - 15 | 1341 | 98 | ||
AK314628.1 | - | 24 - 25 | 2600 | 98 | ||
AL833209.1 | - | 22 - 23 | 2156 | 98 | ||
AL833213.1 | - | 24 - 25 | 2546 | 98 | ||
BC016918.1 | - | 21 - 22 | 2080 | 98 | ||
BC080542.1 | - | 26 - 27 | 2732 | 5479496 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |