Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019580.hg.1 |
ENST00000262623.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019580.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019580.hg.1 |
TC19001448.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019581.hg.1 |
ENST00000262623.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019581.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019581.hg.1 |
TC19001448.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000590916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000590916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000590916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000590916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
ENST00000621873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000590916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000590916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000590916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000590916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
ENST00000621873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010482.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185772.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185772.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185772.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185772.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185772.hg.1 |
TC1900010482.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900185772.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185772.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185772.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185772.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185773.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185773.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185773.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185773.hg.1 |
NM_000704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185773.hg.1 |
TC1900010482.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900185773.hg.1 |
ENST00000262623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185773.hg.1 |
ENST00000592131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185773.hg.1 |
ENST00000592767.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185773.hg.1 |
NM_000704.2 |
Refseq |