Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260598.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000260599.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000429697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000429697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000429697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000429697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000429697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000429697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000429697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000429697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000429697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000429697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000429697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000429697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000464371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000464371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000464371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000464371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000464371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000464371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000464371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000464371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000464371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000464371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000464371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000469936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000469936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000469936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000469936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
ENST00000490823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_000221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007053.hg.1 |
NM_006488.2 |
Refseq |