Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017000534.1 | XP_016856023.1 | 12 - 13 | 2380 | 99 | |
XM_017000535.1 | XP_016856024.1 | 12 - 13 | 2340 | 99 | ||
XM_017000536.1 | XP_016856025.1 | 11 - 12 | 2188 | 99 | ||
GenBank mRNA | AK296788.1 | - | 11 - 12 | 1433 | 99 | |
BC037166.2 | - | 12 - 13 | 2228 | 5090654 | 99 | |
M59371.1 | - | 12 - 13 | 2223 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |