Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011966.hg.1 |
TC03000665.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011966.hg.1 |
ENST00000302192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011966.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011966.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161836.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161836.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161836.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161836.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161836.hg.1 |
TC0300008703.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
ENST00000342480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008703.hg.1 |
NM_015720.3 |
Refseq |