Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032014.2 | NP_114403.1 | 1 - 2 | 87 | 106 | |
GenBank mRNA | BC012167.1 | - | 1 - 2 | 39 | 4637888 | 106 |
BC054865.1 | - | 1 - 2 | 54 | 6456699 | 106 | |
CA413186.1 | - | 3 - 4 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons |