Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006367.hg.1 |
TC13001724.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006367.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006367.hg.1 |
ENST00000379881.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006367.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167597.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167597.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167597.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167597.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167597.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167597.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167597.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167597.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167597.hg.1 |
TC1300010032.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300167576.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167576.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167576.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167576.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167576.hg.1 |
TC1300010031.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
ENST00000511166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010031.hg.1 |
NM_001198910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000317764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
ENST00000379881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_001282147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010032.hg.1 |
NM_017826.2 |
Refseq |