Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003241.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003241.hg.1 |
ENST00000396213.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003241.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003241.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003241.hg.1 |
TC11000243.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003246.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003246.hg.1 |
ENST00000396213.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003246.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003246.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003246.hg.1 |
TC11000243.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003242.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003242.hg.1 |
ENST00000396213.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003242.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003242.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003242.hg.1 |
TC11000243.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003245.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003245.hg.1 |
ENST00000396213.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003245.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003245.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003245.hg.1 |
TC11000243.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003237.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003237.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003237.hg.1 |
TC11000243.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100149393.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149393.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149393.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149393.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149393.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149393.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149393.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149393.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149393.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149393.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149393.hg.1 |
TC1100007006.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100149394.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149394.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149394.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149394.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149394.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149394.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149394.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149394.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149394.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149394.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149394.hg.1 |
TC1100007006.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007006.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149396.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149396.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149396.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149396.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149396.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149396.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149396.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149396.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149396.hg.1 |
TC1100007006.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100149392.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149392.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149392.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149392.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149392.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149392.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149392.hg.1 |
ENST00000396213.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149392.hg.1 |
ENST00000615355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149392.hg.1 |
NM_001144071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149392.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149392.hg.1 |
TC1100007006.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100149387.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149387.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149387.hg.1 |
ENST00000280706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149387.hg.1 |
NM_144972.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149387.hg.1 |
TC1100007006.hg.1 |
Clariom D TC |