Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006722552.3 | XP_006722615.1 | 1 - 2 | 122 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |