Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000369111.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000483993.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000483993.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000483993.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000483993.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000483993.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000483993.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000483993.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000483993.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000483993.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000483993.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000582010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000582010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000582010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000582010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000582010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000582010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000582010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
ENST00000607652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009897.hg.1 |
NM_012113.2 |
Refseq |