Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011687.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011687.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011687.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011687.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011687.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011687.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011687.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011687.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011687.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024877.hg.1 |
ENST00000375366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024877.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024877.hg.1 |
TC17001855.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024876.hg.1 |
ENST00000375366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024876.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024876.hg.1 |
TC17001855.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700194647.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194647.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194647.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194647.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194647.hg.1 |
TC1700011687.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011687.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011687.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011687.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011687.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011687.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011687.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011687.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011687.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011687.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194648.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194648.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194648.hg.1 |
ENST00000375366.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194648.hg.1 |
NM_001080466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194648.hg.1 |
TC1700011687.hg.1 |
Clariom D TC |