Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015255.2 | NP_056070.1 | 28 - 29 | 3362 | 72 | |
XM_005248965.4 | XP_005249022.1 | 28 - 29 | 3479 | 72 | ||
XM_005248966.3 | XP_005249023.1 | 27 - 28 | 2762 | 72 | ||
XM_011514438.2 | XP_011512740.2 | 28 - 29 | 3562 | 72 | ||
XM_011514439.1 | XP_011512741.1 | 19 - 20 | 1929 | 72 | ||
XM_011514440.1 | XP_011512742.1 | 19 - 20 | 1929 | 72 | ||
XM_011514441.2 | XP_011512743.1 | 16 - 17 | 1542 | 72 | ||
XM_017010594.1 | XP_016866083.1 | 28 - 29 | 3563 | 72 | ||
XM_017010595.1 | XP_016866084.1 | 28 - 29 | 3563 | 72 | ||
XM_017010596.1 | XP_016866085.1 | 28 - 29 | 3397 | 72 | ||
XM_017010597.1 | XP_016866086.1 | 28 - 29 | 3564 | 72 | ||
GenBank mRNA | AB002347.1 | - | 16 - 17 | 1624 | 72 | |
AK304070.1 | - | 20 - 21 | 2015 | 72 | ||
AY061884.1 | - | 28 - 29 | 3064 | 72 | ||
BX647467.1 | - | 11 - 12 | 1035 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |