Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004207.hg.1 |
ENST00000299022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004207.hg.1 |
ENST00000356113.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004207.hg.1 |
ENST00000414170.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004207.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004207.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004207.hg.1 |
TC15000442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000560257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000560257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000560257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000560257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000560257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000560664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000560664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000560664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000560664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
ENST00000560664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500152511.hg.1 |
TC1500007365.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000299022.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000356113.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000414170.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000433326.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000559845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000560257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000560257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000560257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000560257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000560257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000560664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000560664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000560664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000560664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
ENST00000560664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007365.hg.1 |
NM_000236.2 |
Refseq |