Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000323084.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000397916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000613245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
ENST00000614657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_001272037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008374.hg.1 |
NM_144991.2 |
Refseq |