Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
ENST00000263398.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
ENST00000278386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
ENST00000352818.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
ENST00000360158.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
ENST00000415148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
ENST00000428726.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
ENST00000433354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
ENST00000433892.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
ENST00000434472.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
ENST00000437706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
ENST00000449691.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
NM_000610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
NM_001001389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
NM_001001390.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
NM_001001391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
NM_001001392.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
NM_001202555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
NM_001202556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
NM_001202557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004284.hg.1 |
TC11000332.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000263398.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000278386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000279452.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000352818.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000415148.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000425428.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000428726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000433892.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000434472.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000442151.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000525211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000526000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000526025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000526669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000527889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000528086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000531118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_000610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001001389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001001390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001001391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001001392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001202555.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001202556.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001202557.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000263398.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000278386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000279452.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000352818.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000415148.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000425428.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000428726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000433892.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000434472.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000442151.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000525211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000526000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000526025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000526669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000527889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000528086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
ENST00000531118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_000610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001001389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001001390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001001391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001001392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001202555.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001202556.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
NM_001202557.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151301.hg.1 |
TC1100007273.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000263398.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000278386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000279452.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000352818.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000415148.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000425428.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000428726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000433892.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000434472.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000442151.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000525211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000526000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000526025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000526669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000527889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000528086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000528455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000531118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_000610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001001389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001001390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001001391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001001392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001202555.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001202556.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001202557.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000263398.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000278386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000279452.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000352818.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000415148.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000425428.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000428726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000433892.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000434472.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000442151.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000525211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000526000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000526025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000526669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000527889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000528086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000528455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
ENST00000531118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_000610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001001389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001001390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001001391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001001392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001202555.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001202556.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
NM_001202557.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151302.hg.1 |
TC1100007273.hg.1 |
Clariom D TC |