Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022637.hg.1 |
TC09001753.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022637.hg.1 |
ENST00000371600.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022637.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022637.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022633.hg.1 |
TC09001753.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022633.hg.1 |
ENST00000371600.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022633.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022633.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000472668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000472668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000472668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000472668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000485589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000485589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000485589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000485589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000485589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000485589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000488145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000488145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000488145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000488145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000488145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000488145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000496498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000496498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000496498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000496498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
ENST00000496498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000472668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000472668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000472668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000472668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000485589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000485589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000485589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000485589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000485589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000485589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000488145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000488145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000488145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000488145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000488145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000488145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000496498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000496498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000496498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000496498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
ENST00000496498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012291.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186937.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186937.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186937.hg.1 |
ENST00000472668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186937.hg.1 |
ENST00000496498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186937.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186937.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186937.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186937.hg.1 |
ENST00000472668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186937.hg.1 |
ENST00000496498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186937.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186937.hg.1 |
TC0900012291.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900186941.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186941.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186941.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186941.hg.1 |
ENST00000371600.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186941.hg.1 |
ENST00000371601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186941.hg.1 |
NM_015392.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186941.hg.1 |
TC0900012291.hg.1 |
Clariom D TC |