Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000490345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000490345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
ENST00000490345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022110.hg.1 |
ENST00000355422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022110.hg.1 |
ENST00000369236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022110.hg.1 |
ENST00000439649.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022110.hg.1 |
ENST00000544592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022110.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022110.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022110.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022110.hg.1 |
TC10001678.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000490345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000490345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
ENST00000490345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011923.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
ENST00000355422.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
ENST00000369234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
ENST00000369236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
ENST00000439649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
NM_001145453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
NM_005264.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
NM_145793.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000181570.hg.1 |
TC1000011923.hg.1 |
Clariom D TC |