Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038372.hg.1 |
TC01002481.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038372.hg.1 |
ENST00000291416.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038372.hg.1 |
ENST00000373430.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038372.hg.1 |
ENST00000373432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038372.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038372.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100201519.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100201519.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100201519.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100201519.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100201519.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100201519.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100201519.hg.1 |
TC0100013647.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |