Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002726.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002726.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002726.hg.1 |
TC20000220.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002725.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002725.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002725.hg.1 |
TC20000220.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002727.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002727.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002727.hg.1 |
TC20000220.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000148846.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148846.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148846.hg.1 |
TC2000007138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148846.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148846.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148847.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148847.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148847.hg.1 |
TC2000007138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148847.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148847.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148849.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148849.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148849.hg.1 |
TC2000007138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148849.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148849.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007138.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148848.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148848.hg.1 |
NM_025227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148848.hg.1 |
TC2000007138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148848.hg.1 |
ENST00000170150.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148848.hg.1 |
NM_025227.2 |
Refseq |