Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004017.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004017.hg.1 |
ENST00000538599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004017.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004017.hg.1 |
TC20000282.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000150578.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150578.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150578.hg.1 |
TC2000007313.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000150578.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150578.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |