Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021214.hg.1 |
TC06001764.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021214.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021214.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000491573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000491573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000491573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
ENST00000491573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000491573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000491573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000491573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
ENST00000491573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011915.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181967.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181967.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181967.hg.1 |
ENST00000244571.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181967.hg.1 |
NM_020745.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181967.hg.1 |
TC0600011915.hg.1 |
Clariom D TC |