Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005013.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005013.hg.1 |
TC10000428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005015.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005015.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005015.hg.1 |
TC10000428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009196.hg.1 |
ENST00000563883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005016.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005016.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005016.hg.1 |
TC10000428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005014.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005014.hg.1 |
TC10000428.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000153771.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153771.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153771.hg.1 |
TC1000007925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000153774.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153774.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153774.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153774.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153774.hg.1 |
TC1000007925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500009196.hg.1 |
ENST00000563883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153772.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153772.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153772.hg.1 |
TC1000007925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000153775.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153775.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153775.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153775.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153775.hg.1 |
TC1000007925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |