Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004725.hg.1 |
ENST00000269187.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004725.hg.1 |
ENST00000440549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004725.hg.1 |
ENST00000543723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004725.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004725.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004725.hg.1 |
TC18000458.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000586829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000586829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000586829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000586829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000586829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000586829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000586829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000586829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000586829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000586829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008474.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
ENST00000586829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
NM_012319.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
TC1800008474.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
ENST00000269187.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
ENST00000440549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
ENST00000586829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
ENST00000590986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
NM_001099406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157831.hg.1 |
NM_012319.3 |
Refseq |