Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012602.hg.1 |
ENST00000291971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012602.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012602.hg.1 |
TC19000914.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008936.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170144.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170144.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170144.hg.1 |
NM_176811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170144.hg.1 |
TC1900008936.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900170144.hg.1 |
ENST00000291971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170144.hg.1 |
ENST00000590542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170144.hg.1 |
NM_176811.2 |
Refseq |