Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012725.hg.1 |
ENST00000301318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012725.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012725.hg.1 |
TC19000926.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000588163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000588163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000588163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000589836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000589836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000589836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000589836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000589836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000594386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000594386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000594386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
ENST00000594386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000588163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000588163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000588163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000589836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000589836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000589836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000589836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000589836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000594386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000594386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000594386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
ENST00000594386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008965.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170509.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170509.hg.1 |
ENST00000589836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170509.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170509.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170509.hg.1 |
NM_020828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170509.hg.1 |
TC1900008965.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900170509.hg.1 |
ENST00000301318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170509.hg.1 |
ENST00000589836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170509.hg.1 |
ENST00000591844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170509.hg.1 |
NM_001308440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170509.hg.1 |
NM_020828.1 |
Refseq |