Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009028.hg.1 |
ENST00000266712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009028.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009028.hg.1 |
TC12000694.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009029.hg.1 |
ENST00000266712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009029.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009029.hg.1 |
TC12000694.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009025.hg.1 |
ENST00000266712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009025.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009025.hg.1 |
TC12000694.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008373.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161548.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161548.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161548.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161548.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161548.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161548.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161548.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161548.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161548.hg.1 |
TC1200008373.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200161547.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161547.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161547.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161547.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161547.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161547.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161547.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161547.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161547.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161547.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161547.hg.1 |
TC1200008373.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200161542.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161542.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161542.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161542.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161542.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161542.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161542.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161542.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161542.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161542.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161542.hg.1 |
TC1200008373.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200161543.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161543.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161543.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161543.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161543.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161543.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161543.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161543.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161543.hg.1 |
TC1200008373.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200161546.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161546.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161546.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161546.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161546.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161546.hg.1 |
ENST00000266712.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161546.hg.1 |
ENST00000547034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161546.hg.1 |
ENST00000549011.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161546.hg.1 |
ENST00000551088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161546.hg.1 |
NM_181783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161546.hg.1 |
TC1200008373.hg.1 |
Clariom D TC |