Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011672.hg.1 |
ENST00000587098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011672.hg.1 |
ENST00000587098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000587019.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000587019.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000587019.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000587019.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000587019.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000589096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000589096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000589096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000589096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000589096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000589669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000589669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000589669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000589669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000589669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000589669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011672.hg.1 |
ENST00000587098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011672.hg.1 |
ENST00000587098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000221554.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000585844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000586666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000587019.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000587019.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000587019.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000587019.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000587019.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000588809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000589096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000589096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000589096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000589096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000589096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000589669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000589669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000589669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000589669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000589669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000589669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
ENST00000593174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011673.hg.1 |
NM_030818.3 |
Refseq |