Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017087.hg.1 |
ENST00000264824.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017087.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017087.hg.1 |
TC19001217.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000590120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000590120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000590974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000590974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000590974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000590974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
ENST00000590974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000590120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000590120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000590974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000590974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000590974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000590974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
ENST00000590974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009765.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179768.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179768.hg.1 |
ENST00000590974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179768.hg.1 |
NM_005583.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179768.hg.1 |
TC1900009765.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900179768.hg.1 |
ENST00000264824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179768.hg.1 |
ENST00000590974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179768.hg.1 |
NM_005583.4 |
Refseq |