Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005914.2 | NP_001005914.1 | 11 - 12 | 1425 | 108 | |
NM_001290060.1 | NP_001276989.1 | 10 - 11 | 1382 | 108 | ||
NM_001290061.1 | NP_001276990.1 | 10 - 11 | 1231 | 108 | ||
NM_001290062.1 | NP_001276991.1 | 4 - 5 | 648 | 108 | ||
NM_001290063.1 | NP_001276992.1 | 4 - 5 | 522 | 108 | ||
NM_004636.3 | NP_004627.1 | 11 - 12 | 1428 | 108 | ||
GenBank mRNA | AB083186.1 | - | 10 - 11 | 1178 | 108 | |
AB209322.1 | - | 9 - 10 | 1608 | 108 | ||
AK293678.1 | - | 4 - 5 | 481 | 108 | ||
BC009113.1 | - | 7 - 8 | 1132 | 4153377 | 108 | |
BC013975.2 | - | 10 - 11 | 1205 | 4121913 | 108 | |
BC024220.2 | - | 10 - 11 | 1205 | 3009807 | 108 | |
BC044602.1 | - | 4 - 5 | 610 | 108 | ||
U28369.1 | - | 10 - 11 | 1371 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |