Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000477763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000477763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
ENST00000477763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000477763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000477763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
ENST00000477763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011807.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
ENST00000477763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
ENST00000369592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
ENST00000369603.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
ENST00000460483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
ENST00000471297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
ENST00000477763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
NM_005871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180908.hg.1 |
TC1000011807.hg.1 |
Clariom D TC |