Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004311.hg.1 |
TC22001422.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004311.hg.1 |
ENST00000331457.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004311.hg.1 |
ENST00000433341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004311.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000469673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000469673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000469673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000469673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000469673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000486584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000486584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000486584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
ENST00000486584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004312.hg.1 |
TC22001422.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004312.hg.1 |
ENST00000331457.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004312.hg.1 |
ENST00000433341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004312.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168628.hg.1 |
TC2200009260.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168628.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168628.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168628.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168628.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168628.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168628.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168627.hg.1 |
TC2200009260.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168627.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168627.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168627.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168627.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168627.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168627.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168627.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168627.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000331457.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000416465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000433341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000469673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000469673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000469673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000469673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000469673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000486584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000486584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000486584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
ENST00000486584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009260.hg.1 |
NM_004147.3 |
Refseq |