Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024449.hg.1 |
ENST00000351960.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024449.hg.1 |
ENST00000358965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024449.hg.1 |
ENST00000359652.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024449.hg.1 |
ENST00000393695.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024449.hg.1 |
ENST00000536544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024449.hg.1 |
NM_006185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024449.hg.1 |
TC11002049.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000351960.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000358965.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000393695.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000541584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000545721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000613205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000616538.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000620566.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
NM_001286561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
NM_006185.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
NR_104476.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000351960.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000358965.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000393695.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000541584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000545721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000613205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000616538.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
ENST00000620566.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
NM_001286561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
NM_006185.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
NR_104476.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188767.hg.1 |
TC1100011539.hg.1 |
Clariom D TC |