Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000360467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000400163.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000400163.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000400163.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000400163.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000400163.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000400163.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000462309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000462309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000462309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000462309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000463084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000463084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000463084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000478681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000478681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000478681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000478681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000480609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000480609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000480609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000480771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000480771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000480771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000480771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000480771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000483683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000483683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000483683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000483683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000483683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000494852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000494852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000494852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000617564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
ENST00000617564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008010.hg.1 |
NM_001846.3 |
Refseq |