Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028338.hg.1 |
TC06002250.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028338.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028338.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194316.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194316.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194316.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194316.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194316.hg.1 |
TC0600013646.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
ENST00000159060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013646.hg.1 |
NM_015718.2 |
Refseq |