Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000524143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000524143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000524143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000524143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000524143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
ENST00000524143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000524143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000524143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000524143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000524143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000524143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
ENST00000524143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008531.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800158574.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158574.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158574.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158574.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158574.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800158574.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158574.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158574.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158574.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158574.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800158574.hg.1 |
TC0800008531.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800158575.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158575.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158575.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158575.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158575.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800158575.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158575.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158575.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158575.hg.1 |
ENST00000520294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158575.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800158575.hg.1 |
TC0800008531.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800158571.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158571.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158571.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158571.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158571.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800158571.hg.1 |
ENST00000220853.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158571.hg.1 |
ENST00000517593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158571.hg.1 |
ENST00000517784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158571.hg.1 |
ENST00000519642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800158571.hg.1 |
NM_014673.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800158571.hg.1 |
TC0800008531.hg.1 |
Clariom D TC |