Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024155.hg.1 |
TC06001961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024155.hg.1 |
ENST00000229971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024155.hg.1 |
ENST00000369244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024155.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024160.hg.1 |
TC06001961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024160.hg.1 |
ENST00000229971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024160.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024156.hg.1 |
TC06001961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024156.hg.1 |
ENST00000229971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024156.hg.1 |
ENST00000369244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024156.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024158.hg.1 |
TC06001961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024158.hg.1 |
ENST00000229971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024158.hg.1 |
ENST00000369244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024158.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187117.hg.1 |
TC0600012647.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187118.hg.1 |
TC0600012647.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012647.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
ENST00000229971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
ENST00000369244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
NM_001278716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
NM_012160.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
NR_103836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
NR_103837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187119.hg.1 |
TC0600012647.hg.1 |
Clariom D TC |