Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012406.hg.1 |
TC01000777.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012406.hg.1 |
ENST00000319942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012406.hg.1 |
ENST00000370824.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012406.hg.1 |
ENST00000496055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012406.hg.1 |
ENST00000535300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012406.hg.1 |
NM_004582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000319942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000461653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000467748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000470201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000471759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000473406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000485459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000489450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000491266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000496055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
NM_004582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
NR_073562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000319942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000461653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000467748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000470201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000471759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000473406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000485459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000489450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000491266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
ENST00000496055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
NM_004582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
NR_073562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238189.hg.1 |
TC0100018237.hg.1 |
Clariom D TC |