Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145814.1 | NP_665813.1 | 3 - 4 | 1134 | 54 | |
GenBank mRNA | AF288386.1 | - | 3 - 4 | 674 | 54 | |
AF361352.1 | - | 3 - 4 | 544 | 54 | ||
BC021552.1 | - | 3 - 4 | 1134 | 3357889 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Mammalian Gene Collection Homo sapiens 1700 Probe spans exons |