Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002641.3 | NP_002632.1 | 5 - 6 | 1308 | 65 | |
NM_020473.3 | NP_065206.3 | 4 - 5 | 531 | 65 | ||
NR_033835.1 | 5 - 6 | 934 | 65 | |||
NR_033836.1 | 5 - 6 | 650 | 65 | |||
XM_011545539.2 | XP_011543841.1 | 4 - 5 | 2132 | 65 | ||
XM_017029581.1 | XP_016885070.1 | 4 - 5 | 1255 | 65 | ||
GenBank mRNA | AK098047.1 | - | 4 - 5 | 744 | 65 | |
AK303538.1 | - | 4 - 5 | 523 | 65 | ||
AK304144.1 | - | 4 - 5 | 1425 | 65 | ||
AK315063.1 | - | 4 - 5 | 1267 | 65 | ||
BC038236.1 | - | 4 - 5 | 1265 | 5733899 | 65 | |
CX870731.1 | - | 4 - 5 | 610 | 65 | ||
D11466.1 | - | 5 - 6 | 1277 | 65 | ||
S74936.1 | - | 3 - 4 | 750 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |