Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003646.hg.1 |
TC09000343.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003646.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003646.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151399.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151399.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151399.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151399.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151399.hg.1 |
TC0900007604.hg.1 |
Clariom D TC |