Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171904.1 | NP_001165375.1 | 19 - 20 | 4067 | 96 | |
NM_014703.2 | NP_055518.1 | 20 - 21 | 4117 | 96 | ||
XM_005276753.4 | XP_005276810.1 | 18 - 19 | 3948 | 96 | ||
XM_005276755.4 | XP_005276812.1 | 19 - 20 | 4029 | 96 | ||
XM_011534273.2 | XP_011532575.1 | 21 - 22 | 4208 | 96 | ||
XM_011534274.2 | XP_011532576.1 | 21 - 22 | 4268 | 96 | ||
XM_011534275.2 | XP_011532577.1 | 21 - 22 | 4183 | 96 | ||
XM_011534276.2 | XP_011532578.1 | 21 - 22 | 4230 | 96 | ||
XM_011534277.2 | XP_011532579.1 | 19 - 20 | 4101 | 96 | ||
XM_017007546.1 | XP_016863035.1 | 20 - 21 | 4126 | 96 | ||
XM_017007547.1 | XP_016863036.1 | 20 - 21 | 4083 | 96 | ||
XM_017007548.1 | XP_016863037.1 | 20 - 21 | 4218 | 96 | ||
XM_017007549.1 | XP_016863038.1 | 21 - 22 | 4268 | 96 | ||
XM_017007550.1 | XP_016863039.1 | 21 - 22 | 4268 | 96 | ||
XM_017007551.1 | XP_016863040.1 | 18 - 19 | 3978 | 96 | ||
GenBank mRNA | AB018343.1 | - | 20 - 21 | 4100 | 96 | |
AF061935.1 | - | 15 - 16 | 3615 | 96 | ||
AL080145.1 | - | 11 - 12 | 2812 | 96 | ||
BC022792.1 | - | 15 - 16 | 2784 | 4853730 | 96 | |
BC110371.1 | - | 19 - 20 | 4057 | 5547856 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |