Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013310.hg.1 |
TC09001012.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013310.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013310.hg.1 |
ENST00000343952.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013310.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013310.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167521.hg.1 |
TC0900009860.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000297991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000463983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000473153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000493581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
ENST00000494313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009860.hg.1 |
NM_004925.4 |
Refseq |