Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009749.hg.1 |
TC06000819.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009749.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009749.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000461673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000461673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
ENST00000461673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161381.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161381.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161381.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161381.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161381.hg.1 |
TC0600008873.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000369278.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000461673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000461673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
ENST00000461673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008873.hg.1 |
NM_015323.4 |
Refseq |