Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016898.hg.1 |
ENST00000225171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016898.hg.1 |
ENST00000339758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016898.hg.1 |
ENST00000483798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016898.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016898.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016898.hg.1 |
TC10001340.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008450.hg.1 |
ENST00000611836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016900.hg.1 |
ENST00000225171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016900.hg.1 |
ENST00000339758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016900.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016900.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016900.hg.1 |
TC10001340.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016896.hg.1 |
ENST00000225171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016896.hg.1 |
ENST00000339758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016896.hg.1 |
ENST00000483798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016896.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016896.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016896.hg.1 |
TC10001340.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016902.hg.1 |
ENST00000225171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016902.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016902.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016902.hg.1 |
TC10001340.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173767.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173767.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173767.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173767.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173767.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173767.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173767.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173767.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173767.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173767.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173767.hg.1 |
TC1000010827.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010827.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173762.hg.1 |
TC1000010827.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173764.hg.1 |
TC1000010827.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000173769.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173769.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173769.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173769.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173769.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173769.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173769.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173769.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173769.hg.1 |
TC1000010827.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008450.hg.1 |
ENST00000611836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
ENST00000225171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
ENST00000339758.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
ENST00000480180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
ENST00000480963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
ENST00000483798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
NM_021800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
NM_201262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173765.hg.1 |
TC1000010827.hg.1 |
Clariom D TC |