Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009940.hg.1 |
ENST00000278886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009940.hg.1 |
ENST00000422516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009940.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009940.hg.1 |
TC20000733.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000336104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000336104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000336104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000336104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000461642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000461642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000461642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000461642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000464285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000464285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000464285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000464285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000464285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000489780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000489780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000489780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000489780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000489780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000496509.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000496509.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000496509.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000496509.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
ENST00000496509.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009939.hg.1 |
ENST00000278886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009939.hg.1 |
ENST00000422516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009939.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009939.hg.1 |
TC20000733.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000161126.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161126.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161126.hg.1 |
ENST00000496509.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161126.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161126.hg.1 |
TC2000008746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000161126.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161126.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161126.hg.1 |
ENST00000496509.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161126.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000278886.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000336104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000336104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000336104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000336104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000422516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000461642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000461642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000461642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000461642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000464285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000464285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000464285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000464285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000464285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000489780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000489780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000489780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000489780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000489780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000496509.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000496509.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000496509.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000496509.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
ENST00000496509.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008746.hg.1 |
NM_025176.5 |
Refseq |