Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000470251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000470251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000470251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000470251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000470251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000470251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000476015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000476015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000476015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000476015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000476015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000486686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000486686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000486686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
ENST00000486686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036370.hg.1 |
TC03002054.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036370.hg.1 |
ENST00000265598.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036370.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036370.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300199333.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300199333.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300199333.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300199333.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300199333.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300199333.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300199333.hg.1 |
TC0300013336.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000265598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000466939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000470251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000470251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000470251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000470251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000470251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000470251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000476015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000476015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000476015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000476015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000476015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000486686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000486686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000486686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
ENST00000486686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013336.hg.1 |
NM_014398.3 |
Refseq |