Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18orf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276342.1 | NP_001263271.1 | 10 - 11 | 1065 | 151 | |
NM_001318707.1 | NP_001305636.1 | 9 - 10 | 980 | 151 | ||
NM_001318708.1 | NP_001305637.1 | 11 - 12 | 1124 | 151 | ||
NM_001318709.1 | NP_001305638.1 | 10 - 11 | 1065 | 151 | ||
NM_013326.4 | NP_037458.3 | 12 - 13 | 1209 | 151 | ||
XM_011525939.1 | XP_011524241.1 | 11 - 12 | 1081 | 151 | ||
XM_017025717.1 | XP_016881206.1 | 10 - 11 | 1018 | 151 | ||
GenBank mRNA | AF143536.1 | - | 11 - 12 | 956 | 151 | |
AK057192.1 | - | 12 - 13 | 1170 | 151 | ||
AK294169.1 | - | 9 - 10 | 927 | 151 | ||
AK295246.1 | - | 11 - 12 | 1081 | 151 | ||
AK296230.1 | - | 10 - 11 | 1016 | 151 | ||
BC002950.1 | - | 12 - 13 | 1134 | 3538017 | 151 | |
BC008305.1 | - | 12 - 13 | 1141 | 3537084 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |